missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-12/IL-23 p40 Homodimer Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 499-ML-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13445224

  • £315.24 / 25µg

Estimated Shipment
10-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Mouse IL-12/IL-23 p40 Homodimer Protein is derived from Sf 21 (baculovirus). The Recombinant Mouse IL-12/IL-23 p40 Homodimer Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
35.8 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse IL-12/IL-23 p40 protein Met23-Ser335
Disulfide-linked homodimer
Yes
16160
IL-12/IL-23 p40 Homodimer
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
Videos
Documents