missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse IL-12/IL-23 p40 Homodimer Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 16160 |
Molecular Weight (g/mol) | 35.8 kDa (monomer) |
Name | IL-12/IL-23 p40 Homodimer |
Quantity | 25μg |
Description
The Recombinant Mouse IL-12/IL-23 p40 Homodimer Protein is derived from Sf 21 (baculovirus). The Recombinant Mouse IL-12/IL-23 p40 Homodimer Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
35.8 kDa (monomer) | |
25μg | |
Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse IL-12/IL-23 p40 protein Met23-Ser335 |
16160 | |
IL-12/IL-23 p40 Homodimer | |
Yes |