Learn More
R&D Systems™ Mouse IL-11 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 418-ML-005
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13434314
  • £332.22 / 5µg
Estimated Shipment date
from Supplier 20-11-2019
Description
Description
The Recombinant Mouse IL-11 Protein is derived from E. coli. The Recombinant Mouse IL-11 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose and with BSA as a carrier protein.
19 kDa
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
Reconstitute at 100 μg/mL in sterile, deionized water.
16156
IL-11
5μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes