missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse IL-11 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 16156 |
Molecular Weight (g/mol) | 19 kDa |
Name | IL-11 |
Recombinant | Yes |
Description
The Recombinant Mouse IL-11 Protein is derived from E. coli. The Recombinant Mouse IL-11 Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
19 kDa | |
Yes |
16156 | |
IL-11 | |
E. coli-derived mouse IL-11 protein Gly23-Leu199,with an N-terminal Met and Pro25-Leu199 |