missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-11 Recombinant Protein

Brand:  R&D Systems™ 418-ML-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13464314

  • £628.45 / 25µg


Estimated Shipment
04-04-2019
Add to basket

Description

Description

Measured in a cell proliferation assay using T11 mouse plasmacytoma cells. Nordan, R.P. et al. (1987) J. Immunol. 139:813. The ED 50 for this effect is 0.04-0.4 ng/mL.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
19 kDa
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
Reconstitute at 100 μg/mL in sterile, deionized water.
16156
IL-11
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, 2 to 8° C under sterile conditions after reconstitution.
Yes
Videos