missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-11 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 418-ML-005/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13444314

  • £342.51 / 5µg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Mouse IL-11 Protein is derived from E. coli. The Recombinant Mouse IL-11 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
19 kDa
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Adipogenesis inhibitory factor, AGIF, AGIFoprelvekin, IL11, IL-11Oprelvekin, interleukin 11, interleukin-11, Oprelvekin
Yes
16156
IL-11
5μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, 2 to 8° C under sterile conditions after reconstitution.
Unlabeled
Reconstitute at 100 μg/mL in sterile, deionized water.
Videos
Documents