missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-10 Recombinant Protein

Brand:  R&D Systems™ 417-ML-005

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13464294

  • £270.00 / 5µg


Estimated Shipment
05-07-2018
Add to basket

Description

Description

Source: E. coli-derived, Ser19-Ser178
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18 kDa
Formulation: Supplied as a 0.2μm filtered solution in Tris and NaCl.
Entrez Gene ID: 16153
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Tris and NaCl with BSA as a carrier protein.
18 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
16153
IL-10
5μg
Noncovalently-linked homodimer
Yes
Videos