missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-1 beta/IL-1F2 Recombinant Protein

Brand:  R&D Systems™ 401-ML-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13462804

  • £838.36 / 25µg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

Source: E. coli-derived, Val118-Ser269, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17 kDa
Formulation: Supplied as a 0.2μm filtered solution in PBS.
Entrez Gene ID: 16176
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
17 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
16176
IL-1 beta/IL-1F2
25μg
Unlabeled
Videos