missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-1 alpha/IL-1F1 Recombinant Protein

Brand:  R&D Systems™ 400-ML-005/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13463964

  • £266.65 / 5µg


Estimated Shipment
03-09-2018
Add to basket

Description

Description

Source: E. coli-derived, Ser6-Ser161
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18 kDa
Formulation: Supplied as a 0.2mm filtered solution in PBS.
Entrez Gene ID: 16175
Regulatory Status: RUO

Specifications

Specifications

Reconstitute at 50μg/mL in sterile PBS.
16175
IL-1 alpha/IL-1F1
5μg
Unlabeled
Lyophilized from a 0.2μm filtered solution in PBS.
18 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
Videos