missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IFN-gamma/IL-4 Dual-Color ELISpot Kit

highly sensitive, microplate-based assays for the detection of cytokine secreting cells

Brand:  R&D Systems™ ELD5217

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13435145

  • £515.97 / 1 set

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Includes:
  • Microplate
  • Biotin-conjugated Detection Antibody
  • HRP-conjugated Detection Antibody
  • Streptavidin-conjugated to Alkaline Phosphatase
  • Dilution Buffers
  • Wash Buffer Concentrate
  • BCIP/NBT Chromogen
  • AEC

Description

Description

Product Features:
  • Detect and quantitate individual cells secreting IFN-γ/IL-4
  • High sensitivity - ELISpot assays can measure responses with frequencies well below 1 in 100,000 cells
  • No in vitro expansion of cells required
  • High-throughput - ELISpot assays use only a small number of primary cells
Specifications

Specifications

Diagnostics
Mouse IFNG/IL-4 Dual-Color ELISpot Kit
One 96-well plate
One 96-well plate coated w/mAb for mouse IFNG and pAb for mouse IL-4, Biotin- and HRP-conjugated detection Abs, Streptavidin-AP, Buffers, BCIP/NBT, AEC, Positive controls
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Videos
Documents