missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse ICAM-2/CD102 Fc Chimera Recombinant Protein

Brand:  R&D Systems™ 774-M2-050

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13486294

  • £246.85 / 50µg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Ser20-Gln222
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 49.5 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Tris, Citric Acid and NaCl.
Entrez Gene ID: 15896
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Tris, Citric Acid and NaCl.
49.5 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
15896
ICAM-2/CD102 Fc Chimera
50μg
Disulfide-linked homodimer
Yes
Videos