missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Hip Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 1568-HP-050

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13439693

  • £424.76 / 50µg

Estimated Shipment
11-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Mouse Hip Protein is derived from NS0. The Recombinant Mouse Hip Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
76 kDa & 73 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse myeloma cell line,NS0-derived mouse Hip protein, (N-terminus) Mouse Hip (Lys24-Arg678 & Asp52-Arg678) Accession &Num; AAD31172 IEGRMDGGGSGGGSGGGS 10-His tag (C-terminus)
Unconjugated
15245
Hip
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos
Documents