missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse Gremlin Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 23892 |
Molecular Weight (g/mol) | 19.6 kDa |
Name | Gremlin |
Quantity | 50μg |
Description
The Recombinant Mouse Gremlin Protein is derived from NS0. The Recombinant Mouse Gremlin Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
19.6 kDa | |
50μg | |
Mouse myeloma cell line,NS0-derived mouse Gremlin protein Lys25-Asp184,with a C-terminal 10-His tag |
23892 | |
Gremlin | |
Yes |