missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse GDF-6/BMP-13 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Gene ID (Entrez) | 242316 |
---|---|
Molecular Weight (g/mol) | 13.7 kDa (monomer) |
Name | GDF-6/BMP-13 |
Quantity | 50μg |
Source | E. coli-derived mouse GDF-6/BMP-13 protein Thr335-Arg454 |
Description
The Recombinant Mouse GDF-6/BMP-13 Protein is derived from E. coli. The Recombinant Mouse GDF-6/BMP-13 Protein has been validated for the following applications: Bioactivity.Specifications
242316 | |
GDF-6/BMP-13 | |
E. coli-derived mouse GDF-6/BMP-13 protein Thr335-Arg454 | |
Disulfide-linked homodimer | |
Recombinant |
13.7 kDa (monomer) | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |