missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse Galectin-7 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 16858 |
Molecular Weight (g/mol) | 15 kDa |
Name | Galectin-7 |
Quantity | 50μg |
Description
The Recombinant Mouse Galectin-7 Protein is derived from E. coli. The Recombinant Mouse Galectin-7 Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
15 kDa | |
50μg | |
E. coli-derived mouse Galectin-7 protein Ser2-Phe136 |
16858 | |
Galectin-7 | |
Yes |