missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Galectin-7 Recombinant Protein

Brand:  R&D Systems™ 1304-GA-050/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459093

  • £298.33 / 50µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: E. coli-derived, Ser2-Phe136
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS and Betamercaptoethanol.
Entrez Gene ID: 16858
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS and Betamercaptoethanol.
15 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
16858
Galectin-7
50μg
Unlabeled
Videos