missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse G-CSF Recombinant Protein

Brand:  R&D Systems™ 414-CS-005

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13464244

  • £161.04 / 5µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: E. coli-derived, Val31-Ala208, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 19 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Citric Acid and Tween™ 20.
Entrez Gene ID: 12985
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Citric Acid and Tween™ 20 with BSA as a carrier protein.
19 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
12985
G-CSF
5μg
Unlabeled
Videos