missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse FGF acidic Recombinant Protein

Brand:  R&D Systems™ 4686-FA-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465034

  • £135.00 / 25µg


Estimated Shipment
29-10-2018
Add to basket

Description

Description

Source: E. coli-derived, Phe16-Asp155, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 16 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT.
Entrez Gene ID: 14164
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT with BSA as a carrier protein.
16 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
14164
FGF acidic
25μg
<0.01 EU per 1μg of the protein by the LAL method.
Yes
Videos