missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse FGF-10 Recombinant Protein

Brand:  R&D Systems™ 6224-FG-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465724

  • £278.00 / 25µg


Estimated Shipment
29-06-2018
Add to basket

Description

Description

Source: E. coli-derived, Ser62-Thr209
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17.1 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT.
Entrez Gene ID: 14165
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT with BSA as a carrier protein.
17.1 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
14165
FGF-10
25μg
Unlabeled
Videos