missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Fas Ligand/TNFSF6 Recombinant Protein

Brand:  R&D Systems™ 526-SA-050/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465374

  • £403.02 / 50µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Pro132-Leu279, with an N-terminal Met and 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 14103
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
18 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
14103
Fas Ligand/TNFSF6
50μg
Unlabeled
Videos