missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse EG-VEGF/PK1 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 246691 |
Molecular Weight (g/mol) | 9.6 kDa |
Name | EG-VEGF/PK1 |
Quantity | 25μg |
Description
The Recombinant Mouse EG-VEGF/PK1 Protein is derived from NS0. The Recombinant Mouse EG-VEGF/PK1 Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
9.6 kDa | |
25μg | |
Mouse myeloma cell line,NS0-derived mouse EG-VEGF/PK1 protein Ala20-Phe105 |
246691 | |
EG-VEGF/PK1 | |
Yes |