missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Draxin Recombinant Protein

Brand:  R&D Systems™ 6149-DR-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465694

  • £347.17 / 25µg


Estimated Shipment
03-09-2018
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Gly26-Ile343, with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 36.4 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 70433
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
36.4 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
70433
Draxin
25μg
Monomer
Yes
Videos