missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CXCL7/Thymus Chemokine-1 aa 48-109 Recombinant Protein

Brand:  R&D Systems™ 1091-CK-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13488463

  • £352.00 / 25µg


Estimated Shipment
02-07-2018
Add to basket

Description

Description

Source: E. coli-derived, Ile48-Ile109
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 6.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 57349
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
6.8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
57349
CXCL7/Thymus Chemokine-1 aa 48-109
25μg
Unlabeled
Videos