missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CXCL3/GRO gamma/CINC-2/DCIP-1 Recombinant Protein

Brand:  R&D Systems™ 5568-CA-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465494

  • £323.41 / 25µg


Estimated Shipment
05-04-2019
Add to basket

Description

Description

Measured by its ability to chemoattract BaF3 mouse pro-B cells transfected with human CXCR2. The ED 50 for this effect is 25-100 ng/mL.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
7.9 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
330122
CXCL3/GRO gamma/CINC-2/DCIP-1
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos