missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CXCL16 Extracellular Domain Recombinant Protein, Asn27-Trp201

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 1165-CX-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13448783

  • £194.04 / 25µg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Mouse CXCL16 (Histidine-tagged) Protein is derived from NS0. The Recombinant Mouse CXCL16 (Histidine-tagged) Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
19.9 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
66102
CXCL16 Extracellular Domain
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos
Documents