missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CXCL10/IP-10/CRG-2 Recombinant Protein

Brand:  R&D Systems™ 466-CR-010/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465024

  • £267.97 / 10µg


Estimated Shipment
04-04-2019
Add to basket

Description

Description

Source: E. coli-derived, Ile22-Pro98, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 15945
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
8.8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
15945
CXCL10/IP-10/CRG-2
10μg
Unlabeled
Videos