missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse CXCL10/IP-10/CRG-2 Recombinant Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 15945 |
Molecular Weight (g/mol) | 8.8 kDa |
Name | CXCL10/IP-10/CRG-2 |
Recombinant | Yes |
Description
The Recombinant Mouse CXCL10/IP-10/CRG-2 Protein is derived from E. coli.Specifications
Unconjugated | |
8.8 kDa | |
Yes |
15945 | |
CXCL10/IP-10/CRG-2 | |
E. coli-derived mouse CXCL10/IP-10/CRG-2 protein Ile22-Pro98,with an N-terminal Met |