missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CX3CL1/Fractalkine Full Length Recombinant Protein

Brand:  R&D Systems™ 472-FF-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465054

  • £298.33 / 25µg


Estimated Shipment
03-04-2019
Add to basket

Description

Description

Measured by its ability to chemoattract freshly isolated human peripheral blood lymphocytes (PBL). The ED 50 for this effect is 0.2-0.6 µg/mL. Measured by its ability to chemoattract BaF3 mouse pro-B cells transfected with mouse CX3CR1. The ED 50 for this effect is 0.03-0.12 µg/mL.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
34 kDa
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Sf 21 (baculovirus)
<0.1 EU per 1μg of the protein by the LAL method.
Yes
20312
CX3CL1/Fractalkine Full Length
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos