missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse CX3CL1/Fractalkine Full Length Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 20312 |
Molecular Weight (g/mol) | 34 kDa |
Name | CX3CL1/Fractalkine Full Length |
Quantity | 25μg |
Description
The Recombinant Mouse CX3CL1/Fractalkine (Full Length) Protein is derived from Sf 21 (baculovirus). The Recombinant Mouse CX3CL1/Fractalkine (Full Length) Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
34 kDa | |
25μg | |
Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse CX3CL1/Fractalkine protein Gln25-Arg337,with a C-terminal 6-His tag |
20312 | |
CX3CL1/Fractalkine Full Length | |
Yes |