missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CX3CL1/Fractalkine aa 25-105 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 571-MF-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13434104

  • £332.22 / 25µg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Mouse CX3CL1/Fractalkine (aa 25-105) Protein is derived from E. coli. The Recombinant Mouse CX3CL1/Fractalkine (aa 25-105) Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
9.0 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
20312
CX3CL1/Fractalkine aa 25-105
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos
Documents