missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CREG Recombinant Protein

Brand:  R&D Systems™ 1697-CR-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458653

  • £298.33 / 25µg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Arg32-Gln220, with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 22.3 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 433375
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
22.3 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
433375
CREG
25μg
Unlabeled
Videos