missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse CREG Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
POA
Specifications
Gene ID (Entrez) | 433375 |
---|---|
Molecular Weight (g/mol) | 22.3 kDa |
Name | CREG |
Quantity | 25μg |
Source | Mouse myeloma cell line,NS0-derived mouse CREG protein Arg32-Gln220,with a C-terminal 6-His tag |
Description
The Recombinant Mouse CREG Protein is derived from NS0. The Recombinant Mouse CREG Protein has been validated for the following applications: Bioactivity.Specifications
433375 | |
CREG | |
Mouse myeloma cell line,NS0-derived mouse CREG protein Arg32-Gln220,with a C-terminal 6-His tag | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
22.3 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |