missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Complement Component C5a Recombinant Protein

Brand:  R&D Systems™ 2150-C5-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460674

  • £309.00 / 25µg


Estimated Shipment
28-06-2018
Add to basket

Description

Description

Source: E. coli-derived, Asn679-Arg755, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 9.0 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 15139
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
9.0 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
15139
Complement Component C5a
25μg
Unlabeled
Videos