missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse Chemerin Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 71660 |
Molecular Weight (g/mol) | 16 kDa |
Name | Chemerin |
Quantity | 25μg |
Description
The Recombinant Mouse Chemerin (aa 17-156) Protein is derived from E. coli. The Recombinant Mouse Chemerin (aa 17-156) Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
16 kDa | |
25μg | |
E. coli-derived mouse Chemerin protein Thr17-Ser156 |
71660 | |
Chemerin | |
Yes |