missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Cerberus 1 Recombinant Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

Brand:  R&D Systems™ 1986-CB-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13440354

  • £347.17 / 25µg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Source: E. coli-derived, Asp18-Pro272, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 28.7 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in HCl.
Entrez Gene ID: 12622
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
28.7 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
12622
Cerberus 1
25μg
Disulfide-linked homodimer
Yes
Videos