missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse CD3+ T Cell Enrichment Column, Small
Designed to prepare purified murine T cell populations via high affinity negative selection
POA
Specifications
Type | Mouse CD3+ T Cell Enrichment Column, Small |
---|---|
Includes | Murine T Cell Selection Columns, 10X Column Buffer Concentrate |
Description
CD3+ is a universal marker for T cells.
Total T cell recovery from these columns ranged between 45% and 65%
Purity (CD3+ cells) of recovered cells ranged between 81% and 88%
Based upon gravitational flow
Simple procedure
No specialized equipment required
Specifications
Mouse CD3+ T Cell Enrichment Column, Small | |
Murine T Cell Selection Columns, 10X Column Buffer Concentrate |