missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CD3+ T Cell Enrichment Column, Small

Designed to prepare purified murine T cell populations via high affinity negative selection

Brand:  R&D Systems™ MTCC-5

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13462226

  • £253.45 / Pack of 5


Estimated Shipment
28-02-2019
Add to basket

Description

Description

CD3+ is a universal marker for T cells.

Total T cell recovery from these columns ranged between 45% and 65%
Purity (CD3+ cells) of recovered cells ranged between 81% and 88%
Based upon gravitational flow
Simple procedure
No specialized equipment required

Specifications

Specifications

Murine T Cell Selection Columns, 10X Column Buffer Concentrate
5/Pk
Mouse CD3+ T Cell Enrichment Column, Small
Videos