missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse CD30 Ligand/TNFSF8 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 21949 |
Molecular Weight (g/mol) | 22 kDa |
Name | CD30 Ligand/TNFSF8 |
Quantity | 50μg |
Description
The Recombinant Mouse CD30 Ligand/TNFSF8 Protein is derived from NS0. The Recombinant Mouse CD30 Ligand/TNFSF8 Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
22 kDa | |
50μg | |
Mouse myeloma cell line,NS0-derived mouse CD30 Ligand/TNFSF8 protein, (N-terminus) MHHHHHHHHHH GGGSGGGSGGGS IEGR Mouse CD30 Ligand (Gln68-Asp239) Accession &Num; P32972 (C-terminus) |
21949 | |
CD30 Ligand/TNFSF8 | |
Yes |