missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse CCL9/10/MIP-1 gamma Truncated, aa 50-122 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 20308 |
Molecular Weight (g/mol) | 8.4 kDa |
Name | CCL9/10/MIP-1 gamma Truncated, aa 50-122 |
Quantity | 25μg |
Description
The Recombinant Mouse CCL9/10/MIP-1 gamma (Truncated) Protein is derived from E. coli. The Recombinant Mouse CCL9/10/MIP-1 gamma (Truncated) Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
8.4 kDa | |
25μg | |
E. coli-derived mouse CCL9/10/MIP-1 gamma protein Gly50-Gln122 |
20308 | |
CCL9/10/MIP-1 gamma Truncated, aa 50-122 | |
Yes |