missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CCL9/10/MIP-1 gamma Truncated, aa 50-122 Recombinant Protein

Brand:  R&D Systems™ 3217-MG-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13462584

  • £212.52 / 25µg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Source: E. coli-derived, Gly50-Gln122
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8.4 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 20308
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
8.4 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
20308
CCL9/10/MIP-1 gamma Truncated, aa 50-122
25μg
Unlabeled
Videos