missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CCL6/C10 Truncated, aa 42-116 Recombinant Protein

Brand:  R&D Systems™ 3210-C-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13462434

  • £298.33 / 25µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: E. coli-derived, Gly42-Ala116
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8.3 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 20305
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
8.3 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
20305
CCL6/C10 Truncated, aa 42-116
25μg
Unlabeled
Videos