missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CCL6/C10 Recombinant Protein

Brand:  R&D Systems™ 487-C-010/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465144

  • £161.04 / 10µg


Estimated Shipment
29-03-2019
Add to basket

Description

Description

Measured by its ability to chemoattract BaF3 mouse pro-B cells transfected with human CCR1. The ED 50 for this effect is 50-250 ng/mL.
Specifications

Specifications

Bioactivity
20305
CCL6/C10
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Lyophilized from a 0.2μm filtered solution in PBS.
10.7 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
Videos