missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CCL3/MIP-1 alpha Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 450-MA-010

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13454854

  • £255.78 / 10µg

Estimated Shipment date
from Supplier 06-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Mouse CCL3/MIP-1 alpha Protein is derived from E. coli. The Recombinant Mouse CCL3/MIP-1 alpha Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
7.8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
C-C motif chemokine 3, MIP1-(a), AI323804, chemokine (C-C motif) ligand 3, G0S19-1, LD78a, LD78alpha, MIP1 alpha, MIP-1 alpha, Mip1a, MIP-1alpha, MIP1-alpha, PAT 464.1, SCYA3, SIS-beta
Yes
20302
CCL3/MIP-1 alpha
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos
Documents