missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CCL24/Eotaxin-2/MPIF-2 aa 27-119 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 1017-MP-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13468303

  • £199.92 / 25µg

Estimated Shipment date
from Supplier 25-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Mouse CCL24/Eotaxin-2/MPIF-2 (aa 27-119) is derived from E. coli. The Recombinant Mouse CCL24/Eotaxin-2/MPIF-2 (aa 27-119) has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
10.3 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
56221
CCL24/Eotaxin-2/MPIF-2 aa 27-119
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos
Documents