missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CCL24/Eotaxin-2/MPIF-2 aa 27-119 Recombinant Protein

Brand:  R&D Systems™ 1017-MP-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458303

  • £174.24 / 25µg


Estimated Shipment
06-03-2019
Add to basket

Description

Description

Source: E. coli-derived, Val27-Val119
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 10.3 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 56221
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
10.3 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
56221
CCL24/Eotaxin-2/MPIF-2 aa 27-119
25μg
Unlabeled
Videos