missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CCL22/MDC Recombinant Protein

Brand:  R&D Systems™ 439-MD-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13464694

  • £245.53 / 25µg


Estimated Shipment
05-04-2019
Add to basket

Description

Description

Measured by its ability to chemoattract BaF3 mouse pro-B cells transfected with human CCR4. The ED 50 for this effect is 0.5-3 ng/mL.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
7.8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
20299
CCL22/MDC
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos