missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CCL21/6Ckine Recombinant Protein

Brand:  R&D Systems™ 457-6C-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13464944

  • £335.29 / 25µg


Estimated Shipment
03-09-2018
Add to basket

Description

Description

Source: E. coli-derived, Ser24-Gly133
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 12 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 18829
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
12 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
18829
CCL21/6Ckine
25μg
Unlabeled
Videos