missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CCL20/MIP-3 alpha Recombinant Protein

Brand:  R&D Systems™ 760-M3-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13486254

  • £223.09 / 25µg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

Source: E. coli-derived, Ala27-Met96
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 7.9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 20297
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
7.9 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
20297
CCL20/MIP-3 alpha
25μg
Unlabeled
Videos