missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CCL2/JE/MCP-1 Recombinant Protein

Brand:  R&D Systems™ 479-JE-010/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465084

  • £225.73 / 10µg


Estimated Shipment
03-04-2019
Add to basket

Description

Description

Measured by its ability to chemoattract BaF3 mouse pro-B cells transfected with human CCR2A. The ED 50 for this effect is 2-10 ng/mL. Measured by its ability to chemoattract 2-day cultured human monocytes. Matsushima, K. et al. (1989) J. Exp. Med. 169:1485. The ED 50 for this effect is 5-20 ng/mL.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
8.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
20296
CCL2/JE/MCP-1
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos