missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CCL17/TARC Recombinant Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 529-TR-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13445384

  • £277.83 / 25µg

Estimated Shipment date
from Supplier 21-10-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Mouse CCL17/TARC Protein is derived from E. coli.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
7.9 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
20295
CCL17/TARC
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos
Documents